Erbium(III) sulfate octahydrate ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
40092-06
40092-09
40092-18
CAAA40092-06EA
77.1
CAD
CAAA40092-06
CAAA40092-09
CAAA40092-18
Erbium(III) sulfate octahydrate ≥99.9% (REO, rare earth oxide basis)
Erbium (III) sulphate octahydrate
Purity: 99.9%
CAS No.: 10031-52-4
Molecular Formula: Er2(SO4)3.8H2O
Formula Weight: 766.82 (622.70anhy)
Physical Form: Crystalline
Density: 3.205
Melting Point: dec.
MDL No.: MFCD00149696
Formula:
Er₂O₁₂S₃·8H₂O MW: 766.82 g/mol (622.70 anhy) Melting Pt: Dec. Density: 3,205 Storage Temperature: Ambient |
MDL Number:
MFCD00149696 CAS Number: 10031-52-4 EINECS: 236-779-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...