Gold(III) acetate ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals

39742-02 39742-03 39742-06
CAAA39742-02EA 252.8 CAD
CAAA39742-02 CAAA39742-03 CAAA39742-06
Gold(III) acetate ≥99.9% (metals basis)
Gold (III) acetate

Purity: 99.9% CAS No.: 15804-32-7 Molecular Formula: Au(O2CCH3)3 Formula Weight: 374.10 Physical Form: Powder

Formula: C₆H₉O₆Yb
Storage Temperature: Ambient
MDL Number: MFCD00210614
CAS Number: 15804-32-7
EINECS: 244-137-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR