Tin (II) trifluoromethanesulfonate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tin trifluoromethanesulfonate, Tin(II) trifluoromethanesulfonate, Tin trifluoromethanesulphonate, Tin triflate, Tin(II) trifluoromethanesulphonate

39603-03 39603-06
CAAA39603-03EA 97.33 CAD
CAAA39603-03 CAAA39603-06
Tin (II) trifluoromethanesulfonate 97%
Tin (II) trifluoromethanesulphonate

Fieser: 11,525 14,306 15,313 17,341 18,357 19,340 20,376 21,424


Purity: 97% CAS No.: 62086-04-8 Molecular Formula: Sn(CF3SO3)2 Formula Weight: 416.82 Physical Form: Powder Sensitivity: Moisture Sensitive UN No.: 3288 MDL No.: MFCD00191251

Formula: C₂F₆O₆S₂Sn
MW: 416.85 g/mol
Melting Pt: Dec.
Storage Temperature: Ambient
MDL Number: MFCD00191251
CAS Number: 62086-04-8
UN: 3288
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR