Bis(cyclopentadienyl)zirconium dichloride ≥99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Zirconocene dichloride

39558-06 39558-14 39558-22
CAAA39558-03EA 57.67 CAD
CAAA39558-03 CAAA39558-06 CAAA39558-14 CAAA39558-22
Bis(cyclopentadienyl)zirconium dichloride ≥99%
Bis(cyclopentadienyl)zirconium dichloride

Fieser: 10,131 15,120 16,121 17,106 18,415 19,414 20,443 21,497


Purity: 99+% CAS No.: 1291-32-3 Molecular Formula: C10H10Cl2Zr Formula Weight: 292.32 Physical Form: Powder, Packaged under argon Sensitivity: Air and Moisture Sensitive Appearance: White to off-white Melting Point: 242-245° Boiling Point: 124-125°/15mm MDL No.: MFCD00003726

Formula: C₁₀H₁₀Cl₂Zr
MW: 292.32 g/mol
Boiling Pt: 124…125 °C (15 mmHg)
Melting Pt: 242…245 °C
Storage Temperature: Ambient
MDL Number: MFCD00003726
CAS Number: 1291-32-3
EINECS: 215-066-8
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR