Tetrachlorobis(tetrahydrofuran)zirconium(IV)
Supplier: Thermo Scientific Chemicals
|
Danger
|
39550-04
39550-09
CAAA39550-04EA
62.25
CAD
CAAA39550-04
CAAA39550-09
Tetrachlorobis(tetrahydrofuran)zirconium(IV)
Tetrachlorobis(tetrahydrofuran)zirconium(IV)
CAS No.: 21959-01-3
Molecular Formula: C8H16Cl4O2Zr
Formula Weight: 377.25
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Appearance: White to off-white
Melting Point: 175-177°
UN No.: 2925
MDL No.: MFCD00145367
Formula:
C₄H₈Cl₄OZr Melting Pt: 175…177 °C Storage Temperature: Ambient |
MDL Number:
MFCD00145367 CAS Number: 21959-01-3 UN: 2925 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...