Lithium trifluoromethanesulfonate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
39322-09
39322-18
CAAA39322-09EA
71.26
CAD
CAAA39322-09
CAAA39322-18
Lithium trifluoromethanesulfonate 97%
Lithium trifluoromethanesulphonate
Purity: 97%
CAS No.: 33454-82-9
Molecular Formula: LiCF3SO3
Formula Weight: 156.01
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: White
Melting Point: 423°
MDL No.: MFCD00013227
Formula:
CF₃LiO₃S Melting Pt: 423 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013227 CAS Number: 33454-82-9 EINECS: 251-528-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...