Mercury(II) trifluoromethanesulfonate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Mercury(II) trifluoromethanesulphonate, Mercury(II) trifluoromethanesulfonate, Mercury triflate

39305-03 39305-06 39305-14
CAAA39305-03EA 77.58 CAD
CAAA39305-03 CAAA39305-06 CAAA39305-14
Mercury(II) trifluoromethanesulfonate 98%
Mercury (II) trifluoromethanesulphonate

Purity: 98% CAS No.: 49540-00-3 Molecular Formula: C2F6HgO6S2 Formula Weight: 498.72 Physical Form: Powder Sensitivity: Moisture Sensitive Appearance: White Melting Point: 350° UN No.: 2025 MDL No.: MFCD00144746

Formula: C₂F₆HgO₆S₂
MW: 498.73
Melting Pt: >350 °C
Storage Temperature: Ambient
MDL Number: MFCD00144746
CAS Number: 49540-00-3
UN: 2025
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR