1,2-Bis(dicyclohexylphosphino)ethane 98%

Supplier: Thermo Scientific Chemicals
Danger

39269-02 39269-04
CAAA39269-02EA 189.6 CAD
CAAA39269-02 CAAA39269-04
1,2-Bis(dicyclohexylphosphino)ethane 98%
1,2-Bis(dicyclohexylphosphino)ethane

Purity: 98% CAS No.: 23743-26-2 Molecular Formula: (C6H11)2PCH2CH2P(C6H11)2 Formula Weight: 422.61 Physical Form: Crystalline, Packaged under argon Sensitivity: Air Sensitive UN No.: 1325 MDL No.: MFCD00015521

Formula: C₂₆H₄₈P₂
MW: 422.607 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00015521
CAS Number: 23743-26-2
UN: 1325
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR