Dicarbonylbis(triphenylphosphine)nickel 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
39242-09
39242-18
39242-27
CAAA39242-09EA
161.16
CAD
CAAA39242-09
CAAA39242-18
CAAA39242-27
Dicarbonylbis(triphenylphosphine)nickel 98%
Dicarbonylbis(triphenylphosphine)nickel
Purity: 98%
CAS No.: 13007-90-4
Molecular Formula: [(C6H5)3P]2Ni(CO)2
Formula Weight: 639.29
Physical Form: Crystalline
Melting Point: 210-215° dec.
UN No.: 3466
MDL No.: MFCD00009591
Formula:
C₃₈H₃₀NiO₂P₂ Melting Pt: 210…215 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00009591 CAS Number: 13007-90-4 EINECS: 235-851-9 ADR: 6.1,II Merck Index: 14,1304 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...