Ammonium tetrachlorocuprate dihydrate
Supplier: Thermo Scientific Chemicals
|
Warning
|
39127-22
39127-36
CAAA39127-22EA
142.67
CAD
CAAA39127-22
CAAA39127-36
Ammonium tetrachlorocuprate dihydrate
Ammonium tetrachlorocuprate dihydrate
Soluble in water, acids, alcohol. Slightly soluble in ammonia
CAS No.: 10060-13-6
Molecular Formula: CuCl2.2NH4Cl.2H2O
Formula Weight: 277.46 (241.43anhy)
Physical Form: Crystals
Density: 1.993
Appearance: Blue
Melting Point: 110° dec.
Solubility: Soluble in water, acids, alcohol. Slightly soluble in ammonia
Merck Reference: 14,514
MDL No.: MFCD00150384
Formula:
H₈Cl₄CuN₂·2H₂O Melting Pt: 110 °C (dec.) Density: 1.993 Storage Temperature: Ambient |
MDL Number:
MFCD00150384 CAS Number: 10060-13-6 EINECS: 239-690-5 Merck Index: 14,514 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...