N,N-Dimethylformamide ≥99.8% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
39117-AP
39117-K2
39117-K7
39117-M1
39117-M6
CAAA39117-APEA
80.94
CAD
CAAA39117-AP
CAAA39117-K2
CAAA39117-K7
CAAA39117-M1
CAAA39117-M6
N,N-Dimethylformamide ≥99.8% ACS
N,N-Dimethylformamide
A solvent for chromogenic substrates
Fieser: 1,273 9,182 11,198 12,203 14,148 16,144 18,146 19,137 21,178
Purity: 99.8+%
CAS No.: 68-12-2
Molecular Formula: HCON(CH3)2
Formula Weight: 73.09
Flash Point: 57°(135°F)
Density: 0.944
Refractive Index: 1.4310
Odor: Amine-like
pH: 7
Melting Point: -61°
Boiling Point: 153°
Beilstein Reference: 605365
Merck Reference: 14,3243
UN No.: 2265
MDL No.: MFCD00003284
A solvent for chromogenic substrates
Formula:
HCON(CH₃)₂ Boiling Pt: 153 °C Melting Pt: –61 °C Density: 0.944 Flash Pt: 57 °C (135 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003284 CAS Number: 68-12-2 EINECS: 200-679-5 UN: 2265 ADR: 3,III REACH: 01-2119475605-32 Merck Index: 14,3243 |
|
Specification Test Results
Appearance | Pass test |
Color (APHA) | Pass test. |
Evaporation residue | 0.005% max. |
Titratable base | 0.003meq/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...