Chromium, plasma standard solution, Specpure®, Cr 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
38728-AD
38728-AE
38728-AP
CAAA38728-ADEA
89.74
CAD
CAAA38728-AD
CAAA38728-AE
CAAA38728-AP
Chromium, plasma standard solution, Specpure®, Cr 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Chromium Standards
Molecular Formula: Cr(NO3)3 in 5% HNO3
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00010944
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...