Titanium, Plasma Standard Solution, Specpure®, Ti 10000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
38725-AD
38725-AE
38725-AP
CAAA38725-ADEA
136.67
CAD
CAAA38725-AD
CAAA38725-AE
CAAA38725-AP
Titanium, Plasma Standard Solution, Specpure®, Ti 10000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Titanium Standards
Molecular Formula: (NH4)2TiF6 in H2O/tr. F-
Physical Form: Liquid
MDL No.: MFCD00011264
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...