Silicon, Plasma Standard Solution, Specpure®, Si 10000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
38723-AD
38723-AE
38723-AP
CAAA38723-ADEA
165.9
CAD
CAAA38723-AD
CAAA38723-AE
CAAA38723-AP
Silicon, Plasma Standard Solution, Specpure®, Si 10000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Silicium Standards
Molecular Formula: (NH4)2SiF6 in H2O/tr. F-
Physical Form: Liquid
MDL No.: MFCD00085311
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...