Chromium, Plasma Standard Solution, Specpure®, Cr 10,000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
38722-AD
38722-AE
38722-AP
CAAA38722-ADEA
113.6
CAD
CAAA38722-AD
CAAA38722-AE
CAAA38722-AP
Chromium, Plasma Standard Solution, Specpure®, Cr 10,000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Chromium Standards
Molecular Formula: Cr(NO3)3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010944
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...