Titanium, Plasma Standard Solution, Specpure®, Ti 1000µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
38720-AD
38720-AE
38720-AP
CAAA38720-ADEA
91.48
CAD
CAAA38720-AD
CAAA38720-AE
CAAA38720-AP
Titanium, Plasma Standard Solution, Specpure®, Ti 1000µg/ml, Thermo Scientific
Standards
Cationic Standards
Titanium Standards
Molecular Formula: (NH4)2TiF6 in H2O/tr. F-
Physical Form: Liquid
MDL No.: MFCD00011264
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...