Molybdenum, Plasma Standard Solution, Specpure®, Mo 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
38719-AD
38719-AE
CAAA38719-ADEA
84.85
CAD
CAAA38719-AD
CAAA38719-AE
CAAA38719-AP
Molybdenum, Plasma Standard Solution, Specpure®, Mo 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Molybdenum Standards
Molecular Formula: (NH4)2MoO4 in 1% NH4OH
Physical Form: Liquid
Odor: Ammonia-like
MDL No.: MFCD00003465
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...