Silicon, Plasma Standard Solution, Specpure®, Si 1000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

38717-AD 38717-AE 38717-AP
CAAA38717-ADEA 99.7 CAD
CAAA38717-AD CAAA38717-AE CAAA38717-AP
Silicon, Plasma Standard Solution, Specpure®, Si 1000 µg/ml, Thermo Scientific
Standards Cationic Standards Silicium Standards

Molecular Formula: (NH4)2SiF6 in H2O/tr. F-
Physical Form: Liquid
MDL No.: MFCD00085311

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR