Methanol-[D₄] (99.8% D)
Supplier: Thermo Scientific Chemicals
|
Danger
|
36501-03
36501-06
36501-14
CAAA36501-03EA
69.52
CAD
CAAA36501-03
CAAA36501-06
CAAA36501-14
Methanol-[D₄] (99.8% D)
Methanol-[D4]
Purity: 99.8%
CAS No.: 811-98-3
Molecular Formula: CD3OD
Formula Weight: 36.07
Physical Form: Liquid
Density: 0.89
Melting Point: -98°
Boiling Point: 64.7°
UN No.: 1230
MDL No.: MFCD00044637
Formula:
D₃COD Boiling Pt: 64.7 °C Melting Pt: –98 °C Density: 0.89 Flash Pt: 11 °C Storage Temperature: Ambient |
MDL Number:
MFCD00044637 CAS Number: 811-98-3 EINECS: 212-378-6 UN: 1230 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...