Cobalt(II) nitrate hexahydrate 98.0-102.0% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cobalt dinitrate hexahydrate, Cobaltous nitrate hexahydrate

36418-22 36418-36
CAAA36418-22EA 113.6 CAD
CAAA36418-22 CAAA36418-36
Cobalt(II) nitrate hexahydrate 98.0-102.0% ACS
Cobalt (II) nitrate hexahydrate

Applications: As a battery additive, in the manufacture of cobalt pigments and invisible inks, for decorating stoneware and procelain, and in the preparation of catalysts
Very soluble in water, alcohol, and most organic solvents


Purity: 98.0-102.0% CAS No.: 10026-22-9 Molecular Formula: Co(NO3)2.6H2O Formula Weight: 291.04 (182.94anhy) Physical Form: Crystalline Density: 1.87 Refractive Index: 1.52 Appearance: Red Odor: Odorless Melting Point: 55-56° Solubility: Very soluble in water, alcohol, and most organic solvents Merck Reference: 14,2444 UN No.: 1477 MDL No.: MFCD00149647


As a battery additive, in the manufacture of cobalt pigments and invisible inks, for decorating stoneware and procelain, and in the preparation of catalysts

Formula: Co(NO₃)₂·6H₂O
MW: 291.03 g/mol
Melting Pt: 55…56 °C
Density: 1.87
Storage Temperature: Ambient
MDL Number: MFCD00149647
CAS Number: 10026-22-9
EINECS: 233-402-1
UN: 1477
ADR: 5.1,III
Merck Index: 14,2444

Order Now

Specification Test Results

Insoluble matter Pass test
Ca Pass test.
Cl 0.002% max.
Cu 0.002% max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR