Aluminon ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
36239-14
36239-22
CAAA36239-14EA
88.64
CAD
CAAA36239-14
CAAA36239-22
Aluminon ACS
Aluminon
CAS No.: 569-58-4
Molecular Formula: C22H23N3O9
Formula Weight: 473.44
Physical Form: Powder
Odor: Odorless
pH: 5.3
Melting Point: 220-225° dec.
Merck Reference: 14,322
MDL No.: MFCD00040925
Formula:
C₂₂H₂₃N₃O₉ Melting Pt: 220…225 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00040925 CAS Number: 569-58-4 EINECS: 209-319-1 Merck Index: 14,322 |
Specification Test Results
Insoluble matter | Pass test |
Residue after ignition | Pass test. |
Sensitivity to aluminum | Pass test. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...