Lithium sulfate monohydrate ≥99.0% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
36216-36
36216-A3
CAAA36216-36EA
244.9
CAD
CAAA36216-36
CAAA36216-A3
Lithium sulfate monohydrate ≥99.0% ACS
Lithium sulphate monohydrate
Soluble in water
Purity: 99.0% min
CAS No.: 10102-25-7
Molecular Formula: Li2SO4.H2O
Formula Weight: 127.96 (109.94anhy)
Physical Form: Crystalline
Density: 2.06
Melting Point: 130° -H2O
Solubility: Soluble in water
Merck Reference: 14,5541
MDL No.: MFCD00149766
Formula:
Li₂SO₄·H₂O MW: 127.96 g/mol Melting Pt: 120 Density: 2.06 Storage Temperature: Ambient |
MDL Number:
MFCD00149766 CAS Number: 10102-25-7 EINECS: 233-820-4 Merck Index: 14,5541 |
Specification Test Results
Loss on drying at 150° | Pass test |
Insoluble matter | Pass test. |
Cl | 0.002% max. |
NO3 | 0.001% max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...