Tungsten, Plasma Standard Solution, Specpure®, W 10000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific
Specpure®
35772-AP 35772-AP 35772-AD 35772-AE 35772-AD 35772-AE
CAAA35772-ADEA 112.81 CAD
AA35772-AE AA35772-AD AA35772-AP CAAA35772-AD CAAA35772-AE CAAA35772-AP
Tungsten, Plasma Standard Solution, Specpure®, W 10000 µg/ml, Thermo Scientific
Standards Cationic Standards Tungsten Standards
Molecular Formula: W in 5% HNO3/tr. HF
Physical Form: Liquid
Odor: Acidic
UN No.: 3264
MDL No.: MFCD00011461
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR