Titanium, AAS Standard Solution, Specpure®, Ti 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
35771-AE
35771-AP
CAAA35771-AEEA
65.1
CAD
CAAA35771-AE
CAAA35771-AP
Titanium, AAS Standard Solution, Specpure®, Ti 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Titanium Standards
Molecular Formula: Ti in 5% HNO3/tr. HF
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011264
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...