Tungsten, Plasma Standard Solution, Specpure®, W 1000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific
Specpure®
35770-AP 35770-AE 35770-AP 35770-AD 35770-AE 35770-AD
CAAA35770-AEEA 115.02 CAD
CAAA35770-AE CAAA35770-AD AA35770-AD AA35770-AE AA35770-AP CAAA35770-AP
Tungsten, Plasma Standard Solution, Specpure®, W 1000 µg/ml, Thermo Scientific
Standards Cationic Standards Tungsten Standards
Molecular Formula: W in 5% HNO3/tr. HF
Physical Form: Liquid
Odor: Acidic
UN No.: 3264
MDL No.: MFCD00011461
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR