Scandium, Plasma Standard Solution, Specpure®, Sc 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
35769-AD
35769-AE
35769-AP
CAAA35769-ADEA
142.52
CAD
CAAA35769-AD
CAAA35769-AE
CAAA35769-AP
Scandium, Plasma Standard Solution, Specpure®, Sc 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Scandium Standards
Molecular Formula: Sc2O3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00016323
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...