Titanium, Plasma Standard Solution, Specpure®, Ti 1000µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
35768-AD
35768-AE
35768-AP
CAAA35768-ADEA
84.85
CAD
CAAA35768-AD
CAAA35768-AE
CAAA35768-AP
Titanium, Plasma Standard Solution, Specpure®, Ti 1000µg/ml, Thermo Scientific
Standards
Cationic Standards
Titanium Standards
Molecular Formula: Ti in 5% HNO3/tr. HF
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011264
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...