Ruthenium, plasma standard solution, Specpure®, Ru 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
35767-AD
35767-AE
35767-AP
CAAA35767-ADEA
156.58
CAD
CAAA35767-AD
CAAA35767-AE
CAAA35767-AP
Ruthenium, plasma standard solution, Specpure®, Ru 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Ruthenium Standards
Molecular Formula: RuCl3 in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011207
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...