Lutetium, Plasma Standard Solution, Specpure®, Lu 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
35765-AD
35765-AE
35765-AP
CAAA35765-ADEA
149.78
CAD
CAAA35765-AD
CAAA35765-AE
CAAA35765-AP
Lutetium, Plasma Standard Solution, Specpure®, Lu 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Lutetium Standards
Molecular Formula: Lu2O3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011098
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...