Titanium, Plasma Standard Solution, Specpure®, Ti 10000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
35759-AD
35759-AE
35759-AP
CAAA35759-ADEA
119.45
CAD
CAAA35759-AD
CAAA35759-AE
CAAA35759-AP
Titanium, Plasma Standard Solution, Specpure®, Ti 10000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Titanium Standards
Molecular Formula: Ti in 5% HNO3/tr. HF
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011264
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...