Scandium, Plasma Standard Solution, Specpure®, Sc 10000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
35755-AD
35755-AE
35755-AP
CAAA35755-ADEA
317.58
CAD
CAAA35755-AD
CAAA35755-AE
CAAA35755-AP
Scandium, Plasma Standard Solution, Specpure®, Sc 10000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Scandium Standards
Molecular Formula: Sc2O3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00016323
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...