Europium, plasma standard solution, Specpure®, Eu 1000µg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

35753-AD 35753-AE 35753-AP
CAAA35753-ADEA 89.11 CAD
CAAA35753-AD CAAA35753-AE CAAA35753-AP
Europium, plasma standard solution, Specpure®, Eu 1000µg/ml, Thermo Scientific
Standards Cationic Standards Europium Standards

Molecular Formula: Eu2O3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010992

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR