Calcium chloride 2,75% (w/v) in aqueous solution

Supplier: Thermo Scientific Chemicals

Synonyms: Chloro calcium, Calcium dichloride, CaCl2

35652-AP 35652-K2
CAAA35652-APEA 52.46 CAD
CAAA35652-AP CAAA35652-K2
Calcium chloride 2,75% (w/v) in aqueous solution
Calcium chloride

Molecular Formula: CaCl2 Formula Weight: 110.99 Physical Form: Liquid MDL No.: MFCD00010903

Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: 1600
Melting Pt: 772
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
Merck Index: 14,01659

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR