Ammonia solution standardised solution 5.0 N
Supplier: Thermo Scientific Chemicals
|
Danger
|
35577-K2
35577-K7
CAAA35577-K2EA
137.46
CAD
CAAA35577-K2
CAAA35577-K7
Ammonia solution standardised solution 5.0 N
Ammonia solution
CAS No.: 1336-21-6
Molecular Formula: H5NO
Formula Weight: 35.05
Physical Form: Liquid
Appearance: Colorless
Odor: Ammonia-like
UN No.: 2672
MDL No.: MFCD00066650
Formula:
NH₃ Storage Temperature: Ambient |
MDL Number:
MFCD00066650 CAS Number: 1336-21-6 EINECS: 215-647-6 UN: 2672 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...