Ammonia solution standardised solution 0.1 N

Supplier: Thermo Scientific Chemicals

35576-K2 35576-K7
CAAA35576-K2EA 100.17 CAD
CAAA35576-K2 CAAA35576-K7
Ammonia solution standardised solution 0.1 N
Ammonia solution

CAS No.: 1336-21-6 Molecular Formula: NH4OH Formula Weight: 35.05 Physical Form: Liquid MDL No.: MFCD00066650

Formula: NH₃
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR