Arsenic, AAS standard solution, Specpure®, As 1000µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
33559-AE
33559-AP
CAAA33559-AEEA
66.52
CAD
CAAA33559-AE
CAAA33559-AP
Arsenic, AAS standard solution, Specpure®, As 1000µg/ml, Thermo Scientific
Standards
Cationic Standards
Arsenic Standards
Molecular Formula: As in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085309
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...