Hexane (mixture of isomers) ≥98.5% ACS

Supplier: Thermo Scientific Chemicals
Danger

33321-K2 33321-K4 33321-K7 33321-M1
CAAA33321-K2EA 97.33 CAD
CAAA33321-K2 CAAA33321-K4 CAAA33321-K7 CAAA33321-M1
Hexane (mixture of isomers) ≥98.5% ACS
Hexane (mixture of isomers)

Purity: 98.5+% CAS No.: 92112-69-1 Molecular Formula: C6H14 Formula Weight: 86.18 Flash Point: -23°(-9°F) Physical Form: Liquid, Mixture of 5 Isomers Density: 0.67 Refractive Index: 1.3795 Appearance: Colorless Melting Point: -95° Boiling Point: 65-70° UN No.: 1208 MDL No.: MFCD02179311

Formula: H₃C(CH₂)₄CH₃
Boiling Pt: 65…70 °C
Melting Pt: –95 °C
Density: 0.67
Flash Pt: <–23 °C (–9 °F)
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 92112-69-1
EINECS: 295-570-2
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Color (APHA) Pass test
Evaporation residue Pass test
Water-soluble titratable acid 0.003meq/g
Sulfur compounds (as S) 0.005% max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR