1,4-Butanediol diacrylate ≥85% (reactive esters) stabilized

Supplier: Thermo Scientific Chemicals
Danger


Stabilization: Stabilised with hydroquinone (HQ) 50-105 ppm

32780-06 32780-14 32780-22
CAAA32780-06EA 45.66 CAD
CAAA32780-06 CAAA32780-14 CAAA32780-22
1,4-Butanediol diacrylate ≥85% (reactive esters) stabilized
1,4-Butanediol diacrylate

Purity: 85+% CAS No.: 1070-70-8 Molecular Formula: C10H14O4 Formula Weight: 198.22 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.051 Refractive Index: 1.4560 Appearance: Colorless Odor: Pleasant Boiling Point: 83°/0.3mm Beilstein Reference: 1777487 UN No.: 1760 MDL No.: MFCD00014940

Formula: C₁₀H₁₄O₄
Boiling Pt: 83 °C (0.3 mmHg)
Density: 1.051
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014940
CAS Number: 1070-70-8
EINECS: 213-979-6
UN: 1760
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR