Dichlorophenylborane ≥96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
32583-03
32583-06
32583-14
CAAA32583-03EA
56.72
CAD
CAAA32583-03
CAAA32583-06
CAAA32583-14
Dichlorophenylborane ≥96%
Dichlorophenylborane
Purity: 96%
CAS No.: 873-51-8
Molecular Formula: C6H5BCl2
Formula Weight: 158.83
Flash Point: 14°(57°F)
Physical Form: Liquid
Density: 1.194
Refractive Index: 1.545
Sensitivity: Moisture Sensitive
Appearance: Colorless
Odor: Acrid
Melting Point: 7°
Boiling Point: 175°
Beilstein Reference: 2243576
UN No.: 2924
MDL No.: MFCD00013612
Formula:
C₆H₅BCl₂ Boiling Pt: 175 °C Melting Pt: 7 °C Density: 1.194 Flash Pt: 14 °C (57 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013612 CAS Number: 873-51-8 UN: 2924 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...