Europium(III) sulfate octahydrate ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
29106-04
29106-09
CAAA29106-04EA
107.6
CAD
CAAA29106-04
CAAA29106-09
Europium(III) sulfate octahydrate ≥99.9% (REO, rare earth oxide basis)
Europium (III) sulphate octahydrate
Purity: 99.9%
CAS No.: 10031-55-7
Molecular Formula: Eu2(SO4)3.8H2O
Formula Weight: 736.23 (592.10anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Merck Reference: 14,3904
MDL No.: MFCD00149704
Formula:
Eu₂O₁₂S₃·8H₂O Melting Pt: 375 °C (-8H₂O) Storage Temperature: Ambient |
MDL Number:
MFCD00149704 CAS Number: 10031-55-7 EINECS: 236-903-3 Merck Index: 14,3904 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...