Specpure® Sulfur, AAS Standard Solution, S 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
22958-AE
22958-AP
CAAA22958-AEEA
62.57
CAD
CAAA22958-AE
CAAA22958-AP
Specpure® Sulfur, AAS Standard Solution, S 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Sulphur Standards
Molecular Formula: (NH4)2SO4 in H2O
Physical Form: Liquid
MDL No.: MFCD00085316
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...