Magnesium trifluoromethanesulfonate
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Magnesium trifluoromethanesulphonate, Magnesium triflate, Magnesium trifluoromethanesulfonate
22340-09
22340-18
CAAA22340-09EA
77.26
CAD
CAAA22340-09
CAAA22340-18
Magnesium trifluoromethanesulfonate
Magnesium trifluoromethanesulphonate
CAS No.: 60871-83-2
Molecular Formula: C2F6MgO6S2
Formula Weight: 322.43
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White
Melting Point:
Formula:
C₂F₆MgO₆S₂ Melting Pt: >310 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042601 CAS Number: 60871-83-2 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...