Tetraethylene glycol dimethyl ether 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,5,8,11,14-Pentaoxapentadecane

20443-30 20443-A1
CAAA20443-30EA 81.53 CAD
CAAA20443-30 CAAA20443-A1
Tetraethylene glycol dimethyl ether 98+%
Tetraethylene glycol dimethyl ether

Purity: 98+% CAS No.: 143-24-8 Molecular Formula: CH3(OCH2CH2)4OCH3 Formula Weight: 222.28 Flash Point: 140°(284°F) Physical Form: Liquid Density: 1.01 Refractive Index: 1.4330 Melting Point: -30° Boiling Point: 275-276° Merck Reference: 14,9208 MDL No.: MFCD00008505

Formula: C₁₀H₂₂O₅
MW: 222.28 g/mol
Boiling Pt: 275…276 °C
Melting Pt: –30 °C
Density: 1.01
Flash Pt: 140 °C (284 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008505
CAS Number: 143-24-8
EINECS: 205-594-7
Merck Index: 14,9208

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR