Potassium trifluoromethanesulfonate 98%

Supplier: Thermo Scientific Chemicals
Warning

20361-09 20361-18
CAAA20361-09EA 81.69 CAD
CAAA20361-09 CAAA20361-18
Potassium trifluoromethanesulfonate 98%
Potassium trifluoromethanesulfonate

Purity: 98% CAS No.: 2926-27-4 Molecular Formula: CF3SO3K Formula Weight: 188.17 Physical Form: Crystalline Sensitivity: Hygroscopic MDL No.: MFCD00042370

Formula: CF₃KO₃S
Storage Temperature: Ambient
MDL Number: MFCD00042370
CAS Number: 2926-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR