Tetrafluoroboric acid diethyl ether complex (50 - 55% HBF₄) (w/w)
Supplier: Thermo Scientific Chemicals
|
Danger
|
18208-AC
18208-AK
CAAA18208-ACEA
152.79
CAD
CAAA18208-AC
CAAA18208-AK
Tetrafluoroboric acid diethyl ether complex (50 - 55% HBF₄) (w/w)
Tetrafluoroboric acid-diethyl ether complex (50-55% tetrafluoroboric acid in diethyl ether)
Reacts in water
CAS No.: 67969-82-8
Molecular Formula: C4H11BF4O
Formula Weight: 161.93
Flash Point: 36°(97°F)
Physical Form: Liquid
Density: 1.19
Appearance: Colorless to pale orange
Solubility: Reacts in water
Merck Reference: 14,4149
UN No.: 2920
MDL No.: MFCD00011345
Formula:
C₄H₁₁BF₄O MW: 161.93 g/mol Density: 1.19 Flash Pt: 36 °C (97 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00011345 CAS Number: 67969-82-8 EINECS: 267-983-8 UN: 2920 ADR: 8,III Merck Index: 14,4149 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...