Ammonium sulfite monohydrate ≥92%
Supplier: Thermo Scientific Chemicals
|
Warning
|
17996-A1
17996-A7
CAAA17996-A1EA
89.11
CAD
CAAA17996-A1
CAAA17996-A7
Ammonium sulfite monohydrate ≥92%
Ammonium sulphite monohydrate
Purity: 92% min
CAS No.: 7783-11-1
Molecular Formula: H8N2O3S.H2O
Formula Weight: 134.15 (116.14anhy)
Physical Form: Crystalline
Density: 1.41
Refractive Index: 1.5150
Sensitivity: Air Sensitive
Appearance: White
Melting Point: 60-70° dec.
Merck Reference: 14,557
MDL No.: MFCD00066094
Formula:
H₈N₂O₃S·1H₂O Melting Pt: 60…70 °C (dec.) Density: 1.41 Storage Temperature: Ambient |
MDL Number:
MFCD00066094 CAS Number: 7783-11-1 EINECS: 233-484-9 Merck Index: 14,557 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...