Trimethylurea
Supplier: Thermo Scientific Chemicals
Synonyms:
1,1,3-Trimethylurea
16475-03
16475-06
16475-14
CAAA16475-03EA
46.45
CAD
CAAA16475-03
CAAA16475-06
CAAA16475-14
Trimethylurea
Trimethylurea
CAS No.: 632-14-4
Molecular Formula: (CH3)2NC(O)NH(CH3)
Formula Weight: 102.14
Physical Form: Solid/semi-solid mass
Sensitivity: Hygroscopic
Odor: Characteristic
MDL No.: MFCD00014868
Formula:
C₄H₁₀N₂O Melting Pt: 74…75 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014868 CAS Number: 632-14-4 EINECS: 211-169-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...