(±)-α-Terpineol ≥96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
16285-22
16285-36
CAAA16285-22EA
90.85
CAD
CAAA16285-22
CAAA16285-36
(±)-α-Terpineol ≥96%
(±)-α-Terpineol
A naturally-occuring monoterpene alcohol
A naturally-occuring monoterpene alcohol
A naturally-occuring monoterpene alcohol
Formula:
C₁₀H₁₈O Boiling Pt: 210…219 °C Melting Pt: 31…32 °C Density: 0.930 Flash Pt: 90 °C (194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001557 CAS Number: 98-55-5 EINECS: 202-680-6 Merck Index: 14,9171 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...