Dipraseodymium tricarbonate octahydrate ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Praseodymium(III) carbonate octahydrate

14609-14 14609-22
CAAA14609-14EA 106.65 CAD
CAAA14609-14 CAAA14609-22
Dipraseodymium tricarbonate octahydrate ≥99.9% (REO, rare earth oxide basis)
Dipraseodymium tricarbonate octahydrate

Purity: 99.9% CAS No.: 14948-62-0 Molecular Formula: Pr2(CO3)3.8H2O Formula Weight: 605.97 (461.85anhy) Physical Form: Crystalline Odor: Odorless MDL No.: MFCD00211307

Formula: C₃O₉Pr₂·8H₂O
MW: 605.97 (461.85 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00211307
CAS Number: 14948-62-0
EINECS: 227-578-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR