Thulium triacetate hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Warning
|
14582-04
14582-09
CAAA14582-04EA
138.88
CAD
CAAA14582-04
CAAA14582-09
CAAA14582-18
Thulium triacetate hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Thulium triacetate hydrate
Purity: 99.9%
CAS No.: 314041-04-8
Molecular Formula: Tm(OOCCH3)3.xH2O
Formula Weight: 346.07(anhy)
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
MDL No.: MFCD00013046
Formula:
C₆H₉O₆Tm·H₂O MW: 346.07 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00013046 CAS Number: 314041-04-8 EINECS: 254-326-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...